Learn More
Acetobromo-alpha-D-glucose, 97%, stab. with 2% calcium carbonate
Description
Acetobromo-α-D-glucose acts as an intermediate in the preparation of beta-glucosides. It is also used as a possible poly(ethylene terephthalate) surface modification reagent to enhance its blood compatibility.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Acetobromo-α-D-glucose acts as an intermediate in the preparation of beta-glucosides. It is also used as a possible poly(ethylene terephthalate) surface modification reagent to enhance its blood compatibility.
Solubility
Soluble in ether, chloroform, ethanol, acetone, dichloromethane, ether, ethyl, methanol, and tetrahydrofuran. Insoluble in water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents, acids, magnesium and aluminum.
Specifications
Specifications
| Melting Point | 84°C to 89°C |
| Quantity | 50 g |
| Beilstein | 96669 |
| Sensitivity | Moisture sensitive |
| Merck Index | 14,60 |
| Solubility Information | Soluble in ether,chloroform,ethanol,acetone,dichloromethane,ether,ethyl,methanol,and tetrahydrofuran. Insoluble in water. |
| Optical Rotation | +198° (c=3 in Chloroform) |
| Formula Weight | 411.22 |
| Percent Purity | 97% |
| Chemical Name or Material | Acetobromo-alpha-D-glucose, stabilized with 2% calcium carbonate |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.