missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetoacetyl CoA synthetase Antibody, mFluor Violet 500 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Acetoacetyl CoA synthetase Polyclonal antibody specifically detects Acetoacetyl CoA synthetase in Human samples. It is validated for ELISA
Specifications
Specifications
| Antigen | Acetoacetyl CoA synthetase |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 500 SE |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | acetoacetate-CoA ligase, acetoacetyl-CoA synthetase, ACSF1FLJ41251, Acyl-CoA synthetase family member 1, EC 6.2.1, EC 6.2.1.16, FLJ12389, homolog of C. elegans supressor of ras 5 (sur-5), Protein sur-5 homolog, SUR-5 |
| Gene ID (Entrez) | 65985 |
| Form | Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?