missing translation for 'onlineSavingsMsg'
Learn More

Acetoacetanilide, 99%

Product Code. 10234510 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
500 g
1 kg
Unit Size:
1kg
500g
5g
This item is not returnable. View return policy

Product Code. 10234510

Brand: Thermo Scientific Acros 102365000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 102-01-2 | C10H11NO2 | 177.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-01-2
Molecular Formula C10H11NO2
Molecular Weight (g/mol) 177.2
MDL Number MFCD00008780
InChI Key DYRDKSSFIWVSNM-UHFFFAOYSA-N
Synonym acetoacetanilide, butanamide, 3-oxo-n-phenyl, acetoacetic anilide, acetoacetamidobenzene, acetylacetanilide, acetoacetylaniline, acetoacetanilid, n-phenylacetoacetamide, acetoacetic acid anilide, n-acetylacetyl aniline
PubChem CID 7592
IUPAC Name 3-oxo-N-phenylbutanamide
SMILES CC(=O)CC(=O)NC1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 83.0°C to 86.0°C
Color White
Flash Point 162°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3COCH2CONHC6H5
Quantity 500 g
Beilstein 12, 518
Merck Index 15, 57
Solubility Information Solubility in water: 5g/L (20 c). Other solubilities: soluble in acids and alkali hydroxide solutions,soluble in alcohol,chloroform,ether,hot benzene,and hot petroleum ether
Formula Weight 177.2
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Acetoacetanilide, 99%
Show More Show Less

RUO – Research Use Only

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.