missing translation for 'onlineSavingsMsg'
Learn More

Acetoacetanilide, 98+%

Product Code. 11428167
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11428167 1000 g 1000g
11438167 250 g 250g
11448167 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11428167 Supplier Thermo Scientific Alfa Aesar Supplier No. A17534.0B
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 102-01-2 | C10H11NO2 | 177.203 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-01-2
Molecular Formula C10H11NO2
Molecular Weight (g/mol) 177.203
MDL Number MFCD00008780
InChI Key DYRDKSSFIWVSNM-UHFFFAOYSA-N
Synonym acetoacetanilide, butanamide, 3-oxo-n-phenyl, acetoacetic anilide, acetoacetamidobenzene, acetylacetanilide, acetoacetylaniline, acetoacetanilid, n-phenylacetoacetamide, acetoacetic acid anilide, n-acetylacetyl aniline
PubChem CID 7592
IUPAC Name 3-oxo-N-phenylbutanamide
SMILES CC(=O)CC(=O)NC1=CC=CC=C1

Specifications

Melting Point 83°C to 86°C
Density 1.26
Boiling Point 129°C (24 mmHg)
Flash Point 150°C (302°F)
Odor Odorless
Quantity 1000 g
Beilstein 473419
Merck Index 14,58
Formula Weight 177.2
Percent Purity ≥98%
Chemical Name or Material Acetoacetanilide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.