missing translation for 'onlineSavingsMsg'
Learn More

Acetoacetamide, TRC

Product Code. 30133515
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133515 50 g 50g
17937574 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133515 Supplier Toronto Research Chemicals Supplier No. TRCA16180550G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5977-14-0
Molecular Formula C4 H7 N O2
Molecular Weight (g/mol) 101.1
Synonym Acetoacetamide (6CI,7CI,8CI), 3-Oxobutanamide, 3-Oxobutyramide, Acetoacetic acid amide, Acetylacetamide, NSC 632231, α-Acetylacetamide
IUPAC Name 3-oxobutanamide
SMILES CC(=O)CC(=O)N

Specifications

Analyte or Component Names Acetoacetamide
Shipping Condition Room Temperature
InChI Formula InChI=1S/C4H7NO2/c1-3(6)2-4(5)7/h2H2,1H3,(H2,5,7)
Solution Type Neat
Industry Type Pharmaceutical
Quantity 50 g
Formula Weight 101.0477
Percent Purity >95%
Chemical Name or Material Acetoacetamide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.