missing translation for 'onlineSavingsMsg'
Learn More

Acetic hydrazide, 96%

Product Code. 11414413
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11414413 500 g 500g
11494403 25 g 25g
11404413 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11414413 Supplier Thermo Scientific Alfa Aesar Supplier No. A10969.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1068-57-1 | C2H6N2O | 74.083 g/mol

Acetic hydrazide is used as an antibiotic for the treatment of mycobacterium tuberculosis. It is also involved in the preparation of sunitinib b reacting with 5-fluoroisatin. Further, it inhibits mycolic acid biosynthesis. It serves as an important intermediate in organic synthesis such as nitric furosemide Long (nifuratrone) in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetic hydrazide is used as an antibiotic for the treatment of mycobacterium tuberculosis. It is also involved in the preparation of sunitinib b reacting with 5-fluoroisatin. Further, it inhibits mycolic acid biosynthesis. It serves as an important intermediate in organic synthesis such as nitric furosemide Long (nifuratrone) in medicine.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1068-57-1
Molecular Formula C2H6N2O
Molecular Weight (g/mol) 74.083
MDL Number MFCD00007610
InChI Key OFLXLNCGODUUOT-UHFFFAOYSA-N
Synonym acethydrazide, acetylhydrazine, acetic hydrazide, acetic acid hydrazide, acetyl hydrazide, monoacetylhydrazine, n-acetylhydrazine, ethanehydrazonic acid, acetic acid, hydrazide, acetyl hydrazine
PubChem CID 14039
ChEBI CHEBI:48978
IUPAC Name acetohydrazide
SMILES CC(=O)NN

Specifications

Melting Point ∼60°C to 68°C
Boiling Point 128°C to 129°C (18 mmHg)
Flash Point >110°C (230°F)
Quantity 500 g
Beilstein 506165
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 74.08
Percent Purity 96%
Chemical Name or Material Acetic hydrazide
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.