missing translation for 'onlineSavingsMsg'
Learn More

Acetic-d3 acid-d, for NMR, 99.5 atom % D, packaged in 0.75 ml ampoules

Product Code. 16348206 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
7.5 mL
Unit Size:
7.50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16348206 7.5 mL 7.50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16348206 Supplier Thermo Scientific Acros Supplier No. 464090075
  • ONLINE EXCLUSIVE PRICE
    £71.00
    £68.90 / 7.5mL
    Save £2.10
    2% Off
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1186-52-3 | C2H4O2 | 64.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Density 1.1300 g/mL
Flash Point 40°C
Refractive Index 1.3709
Specific Gravity 1.13
Quantity 7.5 mL
CAS 1186-52-3
Melting Point 15.0°C to 16.0°C
Boiling Point 115.5°C
Molecular Formula C2H4O2
Linear Formula CD3CO2D
MDL Number MFCD00051051
InChI Key QTBSBXVTEAMEQO-GUEYOVJQSA-N
SMILES [2H]OC(=O)C([2H])([2H])[2H]
IUPAC Name (²H₃)ethan(²H)oic acid
Molecular Weight (g/mol) 64.08
Formula Weight 64.08
Grade NMR
Chemical Name or Material Acetic-d3 acid-d
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetic-d3 acid-d, for NMR, 99.5 atom % D, packaged in 0.75 ml ampoules
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: acetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.