missing translation for 'onlineSavingsMsg'
Learn More

Acetic anhydride, 97+%, ACS reagent

Product Code. p-48003 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
500 mL
1 L
2.5 L
Packaging:
EcoSafPak™ Glass Bottle
Glass bottle
Unit Size:
1 L
2.5 L
500mL
5mL
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.

Product Code. 10731262

Brand: Thermo Scientific Acros 423230050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.

CAS: 108-24-7 | C4H6O3 | 102.089 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Acylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-24-7
Molecular Formula C4H6O3
MDL Number MFCD00008705
InChI Key WFDIJRYMOXRFFG-UHFFFAOYSA-N
Synonym acetic anhydride, acetanhydride, acetic acid, anhydride, acetic oxide, acetyl oxide, ethanoic anhydride, acetyl ether, acetyl anhydride, acetic acid anhydride, anhydride acetique
PubChem CID 7918
ChEBI CHEBI:36610
IUPAC Name acetyl acetate

Specifications

Density 1.087g/mL
Assay Percent Range 97+%
Packaging Glass bottle
Linear Formula (CH3CO)2O
Quantity 5 mL
Beilstein 02, 166
Residue after Evaporation 0.003% max.
Fieser 01,3; 02,7; 05,3; 06,1; 07,1; 08,1; 09,1; 11,1; 12,1; 13,93; 14,150; 15,1
Merck Index 15, 55
SMILES CC(=O)OC(=O)C
Molecular Weight (g/mol) 102.089
Formula Weight 102.09
Percent Purity ≥97.0%
Grade ACS Reagent
Physical Form Liquid
Chemical Name or Material Acetic anhydride, 97+%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Acetic anhydride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.