missing translation for 'onlineSavingsMsg'
Learn More

Acetic Anhydride, Certified AR for Analysis, Fisher Chemical™

Product Code. 11413473 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Unit Size:
2.5 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11413473 500 mL 500mL
10600131 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
Product Code. 11413473 Supplier Fisher Chemical Supplier No. A/0480/PB08
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.

Acetic Anhydride, Certified AR for Analysis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-24-7
Molecular Formula C4H6O3
MDL Number 8705

Specifications

Melting Point -73.1°C
Color Colorless
pH 3
Boiling Point 140°C
Packaging HDPE plastic bottle
Quantity 500 mL
Formula Weight 102.09g/mol
Physical Form Liquid
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Acetic anhydride
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
  • H330-Fatal if inhaled.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.