missing translation for 'onlineSavingsMsg'
Learn More

Acetic anhydride, BAKER ANALYZED™ ACS, J.T. Baker™

Product Code. p-7220831 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10108190 1 L 1 L
10168280 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
Product Code. 10108190 Supplier JT Baker Supplier No. 6004.1000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.

Meets ACS Specifications

Chemical Identifiers

CAS 108-24-7
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.09
MDL Number MFCD00008705
InChI Key WFDIJRYMOXRFFG-UHFFFAOYSA-N
IUPAC Name acetyl acetate
SMILES CC(=O)OC(C)=O

Specifications

Melting Point -73°C
Density 1.08 g/cm³ (20°C)
Boiling Point 139.9°C (1013 hPa)
Flash Point 49°C
Assay Percent Range min. 97.0%
Packaging Glass bottle
Quantity 1 L
Residue after Evaporation max. 0.003%
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.