missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic anhydride, BAKER ANALYZED™ ACS, J.T. Baker™
Shop All J.T.Baker Products
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
Specifications
Specifications
| Melting Point | -73°C |
| Density | 1.08 g/cm³ (20°C) |
| Boiling Point | 139.9°C (1013 hPa) |
| Flash Point | 49°C |
| Assay Percent Range | min. 97.0% |
| Packaging | Glass bottle |
| Quantity | 2.5 L |
| Residue after Evaporation | max. 0.003% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction