Learn More
Acetic anhydride, 99+%, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar L04295.0E
| Quantity | 2500g |
|---|
View more versions of this product
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsAcetic anhydride is most suited for the conversion of cellulose into cellulose acetate and in the manufacture of aspirin. It acts as a wood preservative and solvent in examination of wood fat, fatty and volatile oils and detection of rosins. In many organic reactions it acts as a dehydrating agent. Also used in the production of isoamyl alcohol and 2-methyl furan.
Solubility
Miscible with ether, chloroform and benzene.
Notes
Reacts with water to form acetic acid with the liberation of heat violently. Incompatible with acids, alcohols, bases, oxidizing agents, reducing agents and powdered metals.
Chemical Identifiers
| C4H6O3 | |
| MFCD00008705 | |
| acetic anhydride, acetanhydride, acetic acid, anhydride, acetic oxide, acetyl oxide, ethanoic anhydride, acetyl ether, acetyl anhydride, acetic acid anhydride, anhydride acetique | |
| CHEBI:36610 | |
| CC(=O)OC(=O)C |
Specifications
| 108-24-7 | |
| 138°C to 140°C | |
| Pungent | |
| 1.39 | |
| 2500g | |
| 385737 | |
| 14,56 | |
| Miscible with ether,chloroform and benzene. | |
| CC(=O)OC(=O)C | |
| 102.089 | |
| CHEBI:36610 | |
| ≥99% |
| 1.082 | |
| 49°C (120°F) | |
| C4H6O3 | |
| MFCD00008705 | |
| UN1715 | |
| Moisture sensitive | |
| acetic anhydride, acetanhydride, acetic acid, anhydride, acetic oxide, acetyl oxide, ethanoic anhydride, acetyl ether, acetyl anhydride, acetic acid anhydride, anhydride acetique | |
| WFDIJRYMOXRFFG-UHFFFAOYSA-N | |
| acetyl acetate | |
| 7918 | |
| 102.09 | |
| Acetic anhydride |
Safety and Handling
- Acetic anhydride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H330-Fatal if inhaled.
- EUH071-Corrosive to the respiratory tract.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contain: Acetic anhydride
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.