missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid – Triethylamine Solution 1:1, Honeywell Fluka™

Product Code. 15670900 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15670900 500 mL 500mL
15660900 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15670900 Supplier Honeywell-Fluka Supplier No. 09748500ML

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

for HPLC, 2M:2M concentrate, in H2O

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5204-74-0
Molecular Formula C8H19NO2
Molecular Weight (g/mol) 161.245
MDL Number MFCD00151725
InChI Key AVBGNFCMKJOFIN-UHFFFAOYSA-N
Synonym triethylammonium acetate, teaa, acetic acid, compd. with n,n-diethylethanamine 1:1, acetic acid; n,n-diethylethanamine, ethanamine, n,n-diethyl-, acetate, triethylamine acetate, hoac et3n, acetic acid triethylamine, acetic acid; triethylamine, triethylammonium acetate solution
PubChem CID 165295
IUPAC Name acetic acid;N,N-diethylethanamine
SMILES CCN(CC)CC.CC(=O)O

Specifications

Linear Formula CH3COOH · N(CH2CH3)3
UN Number NONH for all modes of transport
Beilstein 3688333
Formula Weight 161.24g/mol
Density 1.010g/mL at 20°C
Quantity 500 mL
Chemical Name or Material Acetic Acid – Triethylamine Solution 1:1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.