missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic Acid, TraceSELECT™, ≥99%, Honeywell Fluka™
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
Packaging:
Poly Bottle
Description
TraceSELECT™ Ultra and TraceSELECT
- ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
- sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
- the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
- packaged in high-quality containers appropriate for each product
- the reagents are produced and bottled under clean-room conditions
Specifications
Melting Point | 16.2°C |
Boiling Point | 117°C to 118°C |
Color | Colorless |
Appearance | Liquid |
Linear Formula | CH3CO2H |
UN Number | UN2789 |
Beilstein | 506007 |
Formula Weight | 60.05g/mol |
Density | 1.049g/mL at 25°C |
Percent Purity | ≥99% |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'