missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, sodium salt, 99+%, ACS reagent, anhydrous

Product Code. 10155563 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic Bottle
Unit Size:
2.5kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10155563 2.5 kg Plastic Bottle 2.5kg
10351311 500 g Plastic Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10155563 Supplier Thermo Scientific Acros Supplier No. 424250025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-09-3
Molecular Formula C2H3NaO2
Molecular Weight (g/mol) 82.03
MDL Number MFCD00012459
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
Synonym sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
PubChem CID 517045
ChEBI CHEBI:32954
IUPAC Name sodium acetate
SMILES [Na+].CC([O-])=O

Specifications

pH 7.0 to 9.2 (5% soln. at 25°C)
Melting Point 324.0°C
CAS Min % 99.0
CAS Max % 100.0
Color White
Assay Percent Range 99+%
Linear Formula CH3CO2Na
Beilstein 02, 96
Fieser 01,1024; 05,591
Merck Index 15, 8709
Solubility Information Solubility in water: 500g/L (20°C). Other solubilities: slightly soluble in alcohol
Formula Weight 82.03
Percent Purity ≥99%
Grade ACS Reagent
Quantity 2.5 kg
Flash Point >250°C
Infrared Spectrum Authentic
Loss on Drying 1.0% max. (120°C)
Packaging Plastic Bottle
Physical Form Fine Crystalline Powder
Chemical Name or Material Acetic acid, sodium salt
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.