missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, 99.8-100.5%, Honeywell Fluka™
BIOPHARMA

Product Code. 15622900 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 mL
1 L
2.5 L
10 kg
30 kg
4 x 2.5 L
60 kg
6 x 1 L
200 L
Unit Size:
1 L
10kg
2.5 L
200 L
30kg
500mL
60kg
Pack of 4
Pack of 6
This item is not returnable. View return policy

Product Code. 15622900

Brand: Honeywell-Fluka 2722530KG

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss., meets analytical specification of BP, FCC, Ph. Eur., USP, 99.8-100.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym acetasol, acetic acid, glacial, methanecarboxylic acid, vinegar acid, acide acetique, glacial, ethanoic acid, essigsaeure, ethylic acid
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Linear Formula CH3CO2H
UN Number 2789
Formula Weight 60.05g/mol
Percent Purity 99.8 to 100.5%
Grade puriss.
Density 1.049 g/mL at 25°C
Quantity 30 kg
Flash Point 40°C (104°F)
Packaging Plastic Pail
Vapor Density 2.07 (vs. air)
Chemical Name or Material Acetic Acid, Meets analytical specification of Ph. Eur.BP, USP, FCC
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetic acid
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.