missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, Optima™ LC/MS Grade, Fisher Chemical™

Product Code. 11377540 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 x 1 mL Ampule
10 x 1 mL Ampule
50 mL Poly Bottle
Unit Size:
1 ampoule
10 ampoules
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11377540 10 x 1 mL Ampule 10 ampoules
11337540 1 x 1 mL Ampule 1 ampoule
10860701 50 mL Poly Bottle 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 11377540 Supplier Fisher Chemical Supplier No. A11310X1AMP
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

C2H4O2, CAS Number-64-19-7, 50mL, 117 deg.C, Colorless, 60.05g/mol, 16 deg.C, 36152, 2.5, Liquid, 2.1, 1.52kPa at 20 deg.C

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

pH 2.5
Melting Point 16°C
Boiling Point 117°C
Color Colorless
Formula Weight 60.05g/mol
Quantity 10 x 1 mL Ampule
Physical Form Liquid
Vapor Density 2.1
Vapor Pressure 1.52kPa at 20°C
compliance-icons compliance-icons
Product Identifier
  • Acetic acid, Optima LCMS
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Flammable liquid Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.