missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid-octyl ester, Dr. Ehrenstorfer

Product Code. 30323269
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30323269 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30323269 Supplier Dr. Ehrenstorfer Supplier No. DREC10016350
Big Savings for Better Separation
Use promo code 27992
View all promotions
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acetic Acid-Octyl Ester
CAS 112-14-1
Analyte or Component Names Acetic Acid-Octyl Ester
Shipping Condition +20°C
Molecular Formula C10 H20 O2
Certifications/Compliance ISO/IEC 17025
Synonym Acetic Acid-Octyl Ester, Octyl alcohol acetate (6CI), 1-Acetoxyoctane, 1-Octanol acetate, 1-Octyl acetate, Acetic acid octanyl ester, Caprylyl acetate, n-Octyl acetate, NSC 67348, Octanyl acetate, Octyl acetate, Octyl ethanoate
InChI Formula InChI=1S/C10H20O2/c1-3-4-5-6-7-8-9-12-10(2)11/h3-9H2,1-2H3
SMILES CCCCCCCCOC(=O)C
IUPAC Name octyl acetate
Molecular Weight (g/mol) 172.265
Quantity 1 mL
Formula Weight 172.1463 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.