missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, LC/MS grade, Thermo Scientific

Product Code. 30280968 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 mL
10 mL
50 mL
Unit Size:
1mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30280968 1 mL 1mL
30280949 50 mL 50mL
30280957 10 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30280968 Supplier Thermo Scientific Supplier No. T001130001
Big Savings for Better Separation
Use promo code 27992
View all promotions
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

Molecular Weight (g/mol) 60.05 g/mol

Specifications

Quantity 1 mL
CAS 64-19-7
Packaging 1 mL Ampule
Percent Purity 99.70%
Grade LC-MS
Chemical Name or Material Acetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.