missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic Acid Glacial, Pure, Aldehyde Free, Fisher Chemical
Shop All Fisher Chemical Products
Click to view available options
:
500mL, HDPE plastic bottle
Specifications
Specifications
| pH | 2.5 |
| CAS | 64-19-7 |
| Melting Point | 16°C |
| Boiling Point | 117°C |
| Color | Colorless |
| Molecular Formula | C2H4O2 |
| MDL Number | MFCD00036152 |
| InChI Key | QTBSBXVTEAMEQO-UHFFFAOYSA-N |
| SMILES | CC(O)=O |
| IUPAC Name | acetic acid |
| Show More |
Safety and Handling
Product Identifier
- Acetic acid
- Danger
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 A
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction