missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, Glacial, HPLC Grade, J.T. Baker™

Product Code. 15541152 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Clear Glass Bottle
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15541152 2.5 L Clear Glass Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15541152 Supplier Avantor J.T.Baker Supplier No. 951503
  • ONLINE EXCLUSIVE PRICE
    £424.00
    £213.10 / 2.5 L
    Save £210.90
    49% Off
 Restricted Item
Estimated Shipment: 13-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High purity solvents for use in analytical and preparative separations

    For Use in High Performance Liquid Chromatography, Bulk Pharmaceutical Chemical

Specifications

CAS 64-19-7
Molecular Formula C2H4O2
MDL Number MFCD00036152
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
SMILES CC(O)=O
IUPAC Name acetic acid
Molecular Weight (g/mol) 60.05
PubChem CID 176
ChEBI CHEBI:15366
Formula Weight 60.05
Grade Meets USP requirements
Quantity 2.5 L
Flash Point 39.00°C
Packaging Clear Glass Bottle
Specific Gravity 1L = 1.05kg
Chemical Name or Material Acetic Acid, Glacial
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.