missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid Glacial, Certified AR for analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™

Product Code. 12696657 Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 mL
1 L
2.5 L
25 L
Packaging:
HDPE plastic bottle
Plastic Coated Glass bottle
Plastic container
Unit Size:
1 L
2.5 L
25 L
500mL
This item is not returnable. View return policy

Product Code. 12696657

Brand: Fisher Chemical A/0400/PC17

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

C2H4O2, CAS Number-64-19-7, acetasol, acide acetique, essigsaeure, vinegar acid, ethanoic acid, acetic acid, glacial, methanecarboxylic acid, glacial, ethylic acid, 2.5L, 117 deg.C, CHEBI:15366, Colorless, 60.05g/mol, QTBSBXVTEAMEQO-UHFFFAOYSA-N, 16 deg.C, 36152, 60.052, <-0.1ppm

C2H4O2, CAS Number-64-19-7, acetasol, acide acetique, essigsaeure, vinegar acid, ethanoic acid, acetic acid, glacial, methanecarboxylic acid, glacial, ethylic acid, 2.5L, 117 deg.C, CHEBI:15366, Colorless, 60.05g/mol, QTBSBXVTEAMEQO-UHFFFAOYSA-N, 16 deg.C, 36152, 60.052, <-0.1ppm

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

pH 2.5
Melting Point 16°C
Boiling Point 117°C
Color Colorless
Formula Weight 60.05g/mol
Quantity 2.5 L
Packaging Plastic Coated Glass bottle
Physical Form Liquid
Vapor Density 2.1
Vapor Pressure 1.52kPa at 20°C
compliance-icons compliance-icons
Product Identifier
  • Acetic acid
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.