missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, Glacial, BAKER INSTRA-ANALYZED Plus, J.T. Baker™

Product Code. 15538594 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Unit Size:
1 L
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15538594 500 mL 500mL
15548594 1 L 1 L
15558594 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15538594 Supplier JT Baker Supplier No. 9375.0500
  • ONLINE EXCLUSIVE PRICE
    £368.00
    £296.95 / 500mL
    Save £71.05
    19% Off
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Analyzed for up to 64 metals in the low parts-per-billion (ppb) range, ensuring low background interference for conducting critical trace metal analysis.

BAKER INSTRA-ANALYZED™ Plus acids allow for testing of more trace metals with tighter specifications.

  • Quality assured for up to 64 trace metals tested to low parts-per-billion (ppb) levels
  • Lot to lot consistency ensures reliable results between tests
  • Space-saving HDPE packaging with low metallic extractables
  • Enhanced safety with tamper-evident caps
  • Environmentally-friendly recyclable HDPE bottles
  • Strict protocols in an ISO facility ensure quality in manufacturing

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

pH 2.4 (1.0M aq. soln.)
Melting Point 16.4°C
Boiling Point 117.9°C
Color Colorless
Linear Formula CH3CO2H
Solubility Information Miscible in water
Formula Weight 60.05g/mol
Percent Purity 99% (min.)
Grade For use in Trace Metal Analysis
Density 1.05g/mL at 20°C
Quantity 500 mL
Explosion Limit 4 - 19.9 Vol %
Flash Point 39.4°C
Ignition Point 426°C
Odor Pungent, vinegar-like
Packaging HDPE
Physical Form Liquid
Vapor Density 2.1
Vapor Pressure 2.093 kPa
Assay w/w
Chemical Name or Material Acetic acid, glacial
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.