missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic acid, glacial, ACS, 99.7+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
500 mL
2 L
4 x 500 mL
Descrizione
It is used in the production of cellulose acetate for photographic film, polyvinyl acetate for wood glue, synthetic fibers and fabrics. It is used as table vinegar, as preservative and as intermediate in the chemical industry. It is useful for the extraction of glazed ceramic.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
pH | 2.5 |
Melting Point | 16.6°C |
Boiling Point | 118.1°C |
Assay Percent Range | 99.7+% |
UN Number | UN2789 |
Merck Index | 14,55 |
Solubility Information | Miscible with water. |
Formula Weight | 60.05 |
Percent Purity | ≥99.7% |
Grade | ACS |
Vedi altri risultati |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'