missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, glacial, 99+%

Product Code. p-3256833
Click to view available options
Quantity:
250 mL
1 L
4 L
Unit Size:
1 L
250mL
4 L
This item is not returnable. View return policy

Product Code. 11337558

Brand: Thermo Scientific Alfa Aesar 033252.AK

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 64-19-7 | C2H4O2 | 60.05 g/mol

Good polar protic solvent, in its ester form it acts as a solvent for inks paints and coatings. Used in laboratory as a solvent for recrystallization, manufacture of camphor, cellulose acetate, polyvinyl acetate. It involves in Wagner-Meerwein rearrangement. Used for preparation of mono-lithium salt of 5-sulfoisophthalic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Melting Point 16.6°C
Density 1.049
Boiling Point 118.1°C
Flash Point 40°C (104°F)
Assay Percent Range 99+%
Refractive Index 1.3721
Quantity 250 mL
Solubility Information Miscible with water,ethanol,acetone,toluene and immiscible with carbon disulphide.
Formula Weight 60.05
Physical Form Liquid
Chemical Name or Material Acetic acid, glacial
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetic acid, glacial
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Acetic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.