missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, glacial, 99.9985% (metals basis)

Product Code. 11321598
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11321598 100 mL 100mL
11311598 25 mL 25mL
11335819 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11321598 Supplier Thermo Scientific Alfa Aesar Supplier No. 010994.AE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 64-19-7 | C2H4O2 | 60.05 g/mol

Acetic acid is used as a solvent for purification and production of many chemical compounds eg., vinyl acetate monomer. It is a significant chemical reagent and is mainly used in the production of cellulose acetate for photographic film, polyvinyl acetate for wood glue, synthetic fibers and fabrics. It is used as a descaling agent in households. As a food additive, it finds application as a condiment, in the pickling of vegetables and other foods and is also an acidity regulator. In analytical chemistry, for the estimation of weakly alkaline substances such as organic amides, glacial acetic acid is used. Acetic acid is used as antimicrobial preservative or acidulates in a variety of food products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Melting Point 16.6°C
Boiling Point 118.1°C
UN Number UN2789
Merck Index 14,55
Solubility Information Miscible with water,ethanol,acetone,toluene and immiscible with carbon disulphide.
Formula Weight 60.05
Percent Purity 99.9985%
Density 1.049
Quantity 100 mL
Flash Point 40°C (104°F)
Physical Form Liquid
Refractive Index 1.3721
Assay (metals basis)
Chemical Name or Material Acetic Acid, glacial, Glacial
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.