missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, glacial, ACS, 99.7+%

Product Code. 11393358
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 L
4 x 500 mL
Unit Size:
100mL
2 L
2000mL
500mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11393358 100 mL 100mL
11475160 500 mL 500mL
11485160 4 x 500 mL 2000mL
13572310 2 L 2 L
11303368 2 L 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11393358 Supplier Thermo Scientific Alfa Aesar Supplier No. 036289.AE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 64-19-7 | C2H4O2 | 60.05 g/mol

It is used in the production of cellulose acetate for photographic film, polyvinyl acetate for wood glue, synthetic fibers and fabrics. It is used as table vinegar, as preservative and as intermediate in the chemical industry. It is useful for the extraction of glazed ceramic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Melting Point 16.6°C
Boiling Point 118.1°C
Assay Percent Range 99.7+%
UN Number UN2789
Merck Index 14,55
Solubility Information Miscible with water.
Formula Weight 60.05
Percent Purity ≥99.7%
Grade ACS
Density 1.049
Quantity 100 mL
Flash Point 40°C (104°F)
Physical Form Liquid
Refractive Index 1.3721
Chemical Name or Material Acetic Acid, glacial, Glacial
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.