missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid glacial 99-100%, BAKER ANALYZED™ ACS, J.T. Baker™

Product Code. 10371411 Shop All J.T.Baker Products
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Plastic bottle
Unit Size:
1 L
2.5 L
This item is not returnable. View return policy

Product Code. 10371411

Brand: JT Baker 6052.2500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Exceeds ACS Specifications

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Color Max. 10 (APHA)
Assay Percent Range min. 99.7%
Quantity 2.5 L
Packaging Glass bottle
Residue after Evaporation max. 0.001%
compliance-icons compliance-icons
Product Identifier
  • Acetic Acid, Glacial
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contains : acetic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.