missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Acetic acid-d4, 99.5% (Isotopic)

Brand:  Alfa Aesar™ 036469.09

Code : NEW

Additional Details : CAS Number : 1186-52-3 Weight : 0.01000kg Transport : UN number : 2789 Chem class : 1a Pack group : II

Quantity 10g

 View more versions of this product

Product Code. 11367928

  • £51.40 / 10g


Estimated Shipment date
from Supplier 19-02-2018
Add to basket

Specifications

Chemical Identifiers

Acetic acid-d4
Isotopic
MFCD00051051
1186-52-3
64.08
UN2789

Specifications

Acetic acid-d4
Pungent, makes eyes water
118°C
10g
1186-52-3
99.5%
MFCD00051051
1.12g/mL
Moisture Sensitive
17°C
Isotopic
Liquid
64.08
UN2789
Videos
SDS