missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic Acid-d, TRC
Specifications
Specifications
| Chemical Name or Material | Acetic Acid-d |
| CAS | 758-12-3 |
| Molecular Formula | C2 H3 D O2 |
| Quantity | 5 g |
| Synonym | Aci-Jel-d, E 260-d, Ethanoic Acid-d, Ethanoic Acid Monomer-d, Ethylic Acid-d, Glacial Acetic Acid-d, Methanecarboxylic Acid-d, NSC 111201-d, NSC 112209-d, NSC 115870-d, NSC 127175-d, NSC 132953-d, NSC 406306-d, Vinegar Acid-d |
| InChI Formula | InChI=1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)/i/hD |
| SMILES | O=C(C)O[2H] |
| IUPAC Name | deuterio acetate |
| Molecular Weight (g/mol) | 61.06 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?