missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid-d, TRC

Product Code. 30323177
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30323177 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30323177 Supplier Toronto Research Chemicals Supplier No. TRCA16763910G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acetic Acid-d
CAS 758-12-3
Molecular Formula C2 H3 D O2
Quantity 10 g
Synonym Aci-Jel-d, E 260-d, Ethanoic Acid-d, Ethanoic Acid Monomer-d, Ethylic Acid-d, Glacial Acetic Acid-d, Methanecarboxylic Acid-d, NSC 111201-d, NSC 112209-d, NSC 115870-d, NSC 127175-d, NSC 132953-d, NSC 406306-d, Vinegar Acid-d
InChI Formula InChI=1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)/i/hD
SMILES O=C(C)O[2H]
IUPAC Name deuterio acetate
Molecular Weight (g/mol) 61.06
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.