missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, ammonium salt, 97+%, ACS reagent, Thermo Scientific Chemicals

Catalog Number p-45146
Click to view available options
Quantity:
25 g
500 g
2 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
631-61-8
C2H7NO2
77.08
MFCD00013066
USFZMSVCRYTOJT-UHFFFAOYSA-N
ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
517165
CHEBI:62947
azanium;acetate
CC(=O)[O-].[NH4+]
This item is not returnable. View return policy
631-61-8
C2H7NO2
77.08
MFCD00013066
USFZMSVCRYTOJT-UHFFFAOYSA-N
ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
517165
CHEBI:62947
azanium;acetate
CC(=O)[O-].[NH4+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 114.0°C
Color White
CAS Min % 97.0
CAS Max % 100.0
pH 6.7 to 7.3 (5% soln. at 25°C)
Physical Form Adhering Crystals
Quantity 25 g
Assay Percent Range 97+%
Linear Formula CH3CO2NH4
Beilstein 02, 96
Fieser 01,38; 07,11
Flash Point 136°C
Merck Index 15, 492
Solubility Information Solubility in water: 1480g/L (20°C). Other solubilities: soluble in alcohol, slightly soluble in acetone, in methanol: 7.89 g/100mL (15°C)
Formula Weight 77.08
Percent Purity ≥97%
Grade ACS Reagent
Infrared Spectrum Authentic
Packaging Plastic bottle
Residue after Ignition 0.01% max
Density 1.07
Chemical Name or Material Acetic acid, ammonium salt
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'