missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic Acid, 5% vol. Solution, Chem-Lab
Click to view available options
Quantity:
1 L
Packaging:
PE
Unit Size:
1 L
Specifications
Specifications
| CAS | 64-19-7 |
| Molecular Formula | C2H4O2 |
| MDL Number | MFCD00036152 |
| Synonym | ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure |
| InChI Key | QTBSBXVTEAMEQO-UHFFFAOYSA-N |
| SMILES | CC(O)=O |
| Concentration | 50mL CH3COOH / 1L H2O |
| IUPAC Name | acetic acid |
| Molecular Weight (g/mol) | 60.05 |
| PubChem CID | 176 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction