missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, 12%, EP Grade, Reagecon™
BIOPHARMA

Product Code. 11474659 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11474659 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11474659 Supplier Reagecon Supplier No. 1000402

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Manufactured according to European Pharmocopoeia Chapter 4 (4.1.1) and prepared in an ISO 9001 facility. Acetic Acid, 12%, EP Grade, Reagecon™ is a ready-to-use analytical volumetric solution for use in manual or automatic titrations and as a reagent in the production of chemical compounds.

  • Manufactured in an ISO 9001-compliant facility according to Eu. Ph. Chapter 4 (4.1.1) guidelines
  • Provided with a certificate of analysis and a label that shows the lot and expiration date
  • Ready to use reagent eliminates the time and labor of preparing precise volumetric solutions

Specifications

CAS 64-19-7, 7732-18-5
Color Colorless
Molecular Formula C2H4O2
MDL Number MFCD00036152
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
SMILES CC(O)=O
Concentration 12%
IUPAC Name acetic acid
Molecular Weight (g/mol) 60.05
PubChem CID 176
ChEBI CHEBI:15366
Formula Weight 60.05g/mol
Grade Eu. Ph.
Quantity 1 L
Odor Pungent (vinegar)
Physical Form Liquid
Chemical Name or Material Acetic Acid, Dilute
Show More Show Less
compliance-icons
Product Identifier
  • acetic acid, dilute (12%)
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.