missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, 1% v/v aq. soln.

Product Code. 11363667
Change view
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11363667 1 L 1 L
11435020 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11363667 Supplier Thermo Scientific Alfa Aesar Supplier No. 035569.K2
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 64-19-7 | C2H4O2 | 60.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Concentration 1% v/v aq. soln.
Formula Weight 60.05
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Acetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.