missing translation for 'onlineSavingsMsg'
Learn More

Acethydrazide, 95%

Product Code. p-47927 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 10294550

Brand: Thermo Scientific Acros 102350250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1068-57-1 | C2H6N2O | 74.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1068-57-1
Molecular Formula C2H6N2O
Molecular Weight (g/mol) 74.08
MDL Number MFCD00007610
InChI Key OFLXLNCGODUUOT-UHFFFAOYSA-N
Synonym acethydrazide, acetylhydrazine, acetic hydrazide, acetic acid hydrazide, acetyl hydrazide, monoacetylhydrazine, n-acetylhydrazine, ethanehydrazonic acid, acetic acid, hydrazide, acetyl hydrazine
PubChem CID 14039
ChEBI CHEBI:48978
IUPAC Name acetohydrazide
SMILES CC(=O)NN

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 57.0°C to 60.0°C
Color White
Boiling Point 129.0°C (18.0 mmHg)
Flash Point >110°C
Loss on Drying Balance (50°C, 3 Hrs)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (on dry substance)
Packaging Glass bottle
Linear Formula CH3CONHNH2
Quantity 25 g
Beilstein 02,191
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol,acetone,chloroform
Formula Weight 74.08
Percent Purity 95%
Physical Form Moist Crystals and Chunks
Chemical Name or Material Acethydrazide, 95%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acethydrazide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.