missing translation for 'onlineSavingsMsg'
Learn More

Acethydrazide, 95%, Thermo Scientific Chemicals

Catalog Number 10294550
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1068-57-1
C2H6N2O
74.08
MFCD00007610
OFLXLNCGODUUOT-UHFFFAOYSA-N
acethydrazide, acetylhydrazine, acetic hydrazide, acetic acid hydrazide, acetyl hydrazide, monoacetylhydrazine, n-acetylhydrazine, ethanehydrazonic acid, acetic acid, hydrazide, acetyl hydrazine
14039
CHEBI:48978
acetohydrazide
CC(=O)NN
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1068-57-1
C2H6N2O
74.08
MFCD00007610
OFLXLNCGODUUOT-UHFFFAOYSA-N
acethydrazide, acetylhydrazine, acetic hydrazide, acetic acid hydrazide, acetyl hydrazide, monoacetylhydrazine, n-acetylhydrazine, ethanehydrazonic acid, acetic acid, hydrazide, acetyl hydrazine
14039
CHEBI:48978
acetohydrazide
CC(=O)NN

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point 57.0°C to 60.0°C
Color White
Boiling Point 129.0°C (18.0 mmHg)
Flash Point >110°C
Loss on Drying Balance (50°C, 3 Hrs)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (on dry substance)
Packaging Glass bottle
Linear Formula CH3CONHNH2
Quantity 25 g
Beilstein 02,191
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol,acetone,chloroform
Formula Weight 74.08
Percent Purity 95%
Physical Form Moist Crystals and Chunks
Chemical Name or Material Acethydrazide, 95%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acethydrazide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'