missing translation for 'onlineSavingsMsg'
Learn More

Acetazolamide, TRC

Product Code. 30134305
Change view
Click to view available options
Quantity:
50 mg
1 g
100 g
Unit Size:
100g
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134305 100 g 100g
30135592 1 g 1g
17991635 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134305 Supplier Toronto Research Chemicals Supplier No. TRCA161500100G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-66-5
Molecular Formula C4 H6 N4 O3 S2
Molecular Weight (g/mol) 222.25
Synonym Acetazolamide, Acetamide, N-[5-(aminosulfonyl)-1,3,4-thiadiazol-2-yl]-, Acetamide, N-(5-sulfamoyl-1,3,4-thiadiazol-2-yl)- (8CI), N-[5-(Aminosulfonyl)-1,3,4-thiadiazol-2-yl]acetamide, 2-Acetamido-1,3,4-thiadiazole-5-sulfonamide, 2-Acetamido-5-sulfonamido-1,3,4-thiadiazole, 2-Acetylamino-1,3,4-thiadiazole-5-sulfonamide, 5-Acetamide-1,3,4-thiadiazole-2-sulfonamide, 5-Acetamido-1,3,4-thiadiazole-2-sulfonamide, 5-Acetamido-1,3,4-thiodiazole-2-sulfonamide, 5-Acetylamino-1,3,4-thiadiazol-2-ylsulfonamide, AZM, Acetamox, Acetazolamid, Atenezol, Carbonic anhydrase inhibitor 6063, Cidamex, Defiltran, Dehydratin, Diacarb, Diakarb, Diamox, Didoc, Diluran, Diuramid, Diuramide, Diureticum-Holzinger, Diuriwas, Diutazol, Donmox, Edemox, Eumicton, Fonurit, Glaupax, Glupax, L 579486, N-(5-Sulfamoyl-1,3,4-thiadiazol-2-yl)acetamide, N-(5-Sulfamoyl-1,3,4-thiadiazol-2-yl)acetamide, NSC 145177, Natrionex, Nephramid, Nephramide, Phonurit
IUPAC Name N-(5-sulfamoyl-1,3,4-thiadiazol-2-yl)acetamide
SMILES CC(=O)Nc1nnc(s1)S(=O)(=O)N

Specifications

Quantity 100 g
InChI Formula InChI=1S/C4H6N4O3S2/c1-2(9)6-3-7-8-4(12-3)13(5,10)11/h1H3,(H2,5,10,11)(H,6,7,9)
Formula Weight 221.9881
Percent Purity >95%
Chemical Name or Material Acetazolamide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.