missing translation for 'onlineSavingsMsg'
Learn More

Acetazolamide, MedChemExpress

Product Code. 16481058
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481058 5 g 5g
16402279 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16481058 Supplier Medchem Express Supplier No. HYB0782
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Acetazolamide is a carbonic anhydrase (CA) IX inhibitor with an IC50 of 30 nM for hCA IX. Acetazolamide has diuretic, antihypertensive and anti-gonococcal activities.

Acetazolamide is a carbonic anhydrase (CA) IX inhibitor with an IC50 of 30 nM for hCA IX. Acetazolamide has diuretic, antihypertensive and anti-gonococcal activities.

Specifications

Chemical Name or Material Acetazolamide
CAS 59-66-5
Color White
Purity Grade Notes Research
Molecular Formula C4H6N4O3S2
Quantity 5 g
Solubility Information DMSO : 50 mg/mL (224.97 mM; Need ultrasonic) ∣H2O : 1 mg/mL (4.50 mM; Need ultrasonic)
SMILES CC(NC1=NN=C(S(=O)(N)=O)S1)=O
Molecular Weight (g/mol) 222.25
Formula Weight 222.25
Percent Purity 99.63%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.