missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Acetazolamide
GREENER_CHOICE

Product Code. 18779516
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18779516 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18779516 Supplier Tocris Bioscience™ Supplier No. 6742/100
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Carbonic anhydrase inhibitor

Acetazolamide is a carbonic anhydrase inhibitor. Modulates epileptiform synchronization in rat brain slices. Increases speed of synaptic vesicle recycling and changes synaptic performance in ex vivo murine neuromuscular junction. Lowers intracranial pressure in healthy rats. Also potentiates the antitumor effects of the HDAC inhibitor MS 275 (Cat. No. 6208).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[5-(Aminosulfonyl)-1,3,4-thiadiazol-2-yl]acetamide
CAS 59-66-5
Quantity 100 mg
Molecular Formula C4H6N4O3S2
Formula Weight 222.25
Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.