missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACETATE BUFFER SOLUTION PH 4.65, SODIUM ACETATE / ACETIC ACID, Honeywell Fluka™
Shop All Honeywell Fluka ProductsDescription
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Chemical Name or Material | ACETATE BUFFER SOLUTION PH 4.65 |
| Name Note | SODIUM ACETATE / ACETIC ACID |
| CAS | 127-09-3, 64-19-7, 7732-18-5 |
| CAS Min % | ≥90.0000% |
| CAS Max % | ≤100.0000%, <1.0000% |
| Density | 1 g/cm3 |
| Quantity | 6 x 1 L |
| Solution Type | Buffer |
| Linear Formula | HNO3 |
| MDL Number | MFCD00011349 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?