missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DRI™ Acetaminophen Serum Toxicology Assay

Product Code. 17247326 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 x 8 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17247326 25 x 8 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 17247326 Supplier Thermo Scientific™ Supplier No. 1086
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Identify the overdose class of drug in emergency room situations where urine samples are not readily available with this Acetaminophen serum toxicology assay.

Identify the overdose class of drug in emergency room situations where urine samples are not readily available with Thermo Scientific™ DRI™ Acetaminophen Serum Toxicology Assay. Provided in convenient packaging with separate calibrators, these liquid, ready-to-use assays have the flexibility to run on a variety of clinical chemistry analyzers, providing objective results.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description Acetaminophen Assay
Detectable Analytes Acetaminophen
Sample Type Serum
Type Reagent
Quantity 25 x 8 mL
compliance-icons
Product Identifier
  • DRI acetominophem serum assay
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P363-Wash contaminated clothing before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Tris-Hydrochloride,Tromethamine, Bovineserum albumin, Sodium azide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.