missing translation for 'onlineSavingsMsg'
Learn More

Acetaminophen metabolite 3-hydroxy-acetaminophen, MedChemExpress

Product Code. 16478926
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16478926 100 mg 100mg
16441218 5 mg 5mg
16438788 10 mg 10mg
16414707 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16478926 Supplier Medchem Express Supplier No. HYG0004
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 3-hydroxy-acetaminophen is a metabolite of Acetaminophen, which is a pain medicine.

3-hydroxy-acetaminophen is a metabolite of Acetaminophen, which is a pain medicine.

Specifications

Chemical Name or Material Acetaminophen metabolite 3-hydroxy-acetaminophen
CAS 37519-14-5
Color Gray
Purity Grade Notes Research
Molecular Formula C8H9NO3
Quantity 100 mg
Synonym 3-Hydroxyacetaminophen
Solubility Information DMSO : 250 mg/mL (1495.57 mM; Need ultrasonic)
SMILES OC1=C(O)C=C(NC(C)=O)C=C1
Molecular Weight (g/mol) 167.16
Formula Weight 167.16
Percent Purity 99.03%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.