missing translation for 'onlineSavingsMsg'
Learn More

Acetaminophen, MedChemExpress

MedChemExpress Acetaminophen (Paracetamol) is a selective cyclooxygenase-2 (COX-2) inhibitor with an IC50 of 25.8 μM; is a widely used antipyretic and analgesic agent. Acetaminophen is a potent hepatic N-acetyltransferase 2 (NAT2) inhibitor.

£72.60 - £114.95

Specifications

Chemical Name or Material Acetaminophen
CAS 103-90-2
Color White
Purity Grade Notes Research
Molecular Formula C8H9NO2
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16440618
Encompass_Preferred
View Documents
Medchem Express
HY-66005
5 g
£72.60
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16408168
Encompass_Preferred
View Documents
Medchem Express
HY-66005
10 g
£114.95
10g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Acetaminophen (Paracetamol) is a selective cyclooxygenase-2 (COX-2) inhibitor with an IC50 of 25.8 μM
Specifications

Specifications

Acetaminophen
White
C8H9NO2
DMSO : 250 mg/mL (1653.88 mM; Need ultrasonic) ∣H2O : 10 mg/mL (66.16 mM; Need ultrasonic)
151.16
98.79%
Powder
103-90-2
Research
Paracetamol 4-Acetamidophenol 4'-Hydroxyacetanilide
O=C(C)NC1=CC=C(O)C=C1
151.16
Research
COVID-19-immunoregulation
Videos
Safety and Handling

Safety and Handling

H302∣H315∣H319∣H335

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣The compound is unstable in solutions, freshly prepared is recommended.

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣The compound is unstable in solutions, freshly prepared is recommended.

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.