missing translation for 'onlineSavingsMsg'
Learn More

Acetaminophen Acetate (Acetaminophen Impurity), TRC

Product Code. 30135292
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135292 1 g 1g
17917114 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135292 Supplier Toronto Research Chemicals Supplier No. TRCA1612151G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2623-33-8
Molecular Formula C10 H11 N O3
Molecular Weight (g/mol) 193.2
Synonym 4-(Acetylamino)phenyl acetate, Paracetamol Imp. H (EP), N,O-Diacetyl-4-aminophenol, Acetamide, N-[4-(acetyloxy)phenyl]-, Acetanilide, 4'-hydroxy-, acetate (6CI,7CI), Acetanilide, 4'-hydroxy-, acetate (ester) (8CI), N-[4-(Acetyloxy)phenyl]acetamide, 4-Acetamidophenyl acetate, 4-Acetylaminophenyl acetate, 4'-Acetoxyacetanilide, Acetaminophen acetate, Diacetamate, N-(4-Acetoxyphenyl)acetamide, N-Acetyl-4-aminophenyl acetate, NSC 33893, NSC 6083, O-Acetylparacetamol, p-Acetamidophenyl acetate, p-Acetoxyacetanilide
IUPAC Name (4-acetamidophenyl) acetate
SMILES CC(=O)Nc1ccc(OC(=O)C)cc1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C10H11NO3/c1-7(12)11-9-3-5-10(6-4-9)14-8(2)13/h3-6H,1-2H3,(H,11,12)
Formula Weight 193.0739
Chemical Name or Material 4-(Acetylamino)phenyl Acetate (N,O-Diacetyl-4-aminophenol)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.