missing translation for 'onlineSavingsMsg'
Learn More

Acetamide, 99%

Product Code. 11464304
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11464304 1000 g 1000g
11474304 250 g 250g
11484304 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11464304 Supplier Thermo Scientific Alfa Aesar Supplier No. A12589.0B
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 60-35-5 | C2H5NO | 59.068 g/mol

Acetamide is used as an industrial solvent, plasticizer, wetting and penetrating agent. It is also used for the transamidation of carboxamides in 1,4-dioxane. It finds application in lacquers and soldering flux. Further, it acts as a precursor to thioacetamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetamide is used as an industrial solvent, plasticizer, wetting and penetrating agent. It is also used for the transamidation of carboxamides in 1,4-dioxane. It finds application in lacquers and soldering flux. Further, it acts as a precursor to thioacetamide.

Solubility
Soluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids, strong bases and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-35-5
Molecular Formula C2H5NO
Molecular Weight (g/mol) 59.068
MDL Number MFCD00008023
InChI Key DLFVBJFMPXGRIB-UHFFFAOYSA-N
Synonym ethanamide, acetic acid amide, methanecarboxamide, acetimidic acid, amide c2, ethanimidic acid, acetamid, amid kyseliny octove, caswell no. 003h, acetimidic acid van
PubChem CID 178
ChEBI CHEBI:49028
IUPAC Name acetamide
SMILES CC(=O)N

Specifications

Melting Point 76°C to 81°C
Density 1.159
Boiling Point 220°C to 222°C
Quantity 1000 g
Beilstein 1071207
Sensitivity Hygroscopic
Merck Index 14,43
Solubility Information Soluble in water.
Formula Weight 59.07
Percent Purity 99%
Chemical Name or Material Acetamide
Show More Show Less
compliance-icons
Product Identifier
  • Acetamide
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Acetamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.