missing translation for 'onlineSavingsMsg'
Learn More

Acetamide, 99%, for analysis

Product Code. 15236028 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15236028 250 g 250g
15226028 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15236028 Supplier Thermo Scientific Acros Supplier No. 451872500

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 60-35-5 | C2H5NO | 59.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-35-5
Molecular Formula C2H5NO
Molecular Weight (g/mol) 59.06
InChI Key DLFVBJFMPXGRIB-UHFFFAOYSA-N
Synonym ethanamide, acetic acid amide, methanecarboxamide, acetimidic acid, amide c2, ethanimidic acid, acetamid, amid kyseliny octove, caswell no. 003h, acetimidic acid van
PubChem CID 178
ChEBI CHEBI:49028
IUPAC Name acetamide
SMILES CC(=O)N

Specifications

Melting Point 79.0°C to 81.0°C
Boiling Point 221.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Linear Formula CH3CONH2
Quantity 250 g
Merck Index 15,44
Solubility Information Solubility in water: 2000g/L (20°C). Other solubilities: 1 g/2 mL alcohol - 1 g/6 mL pyridine, soluble in chloroform, glycerol and hot benzene, slightly soluble in ether
Formula Weight 59.06
Percent Purity 99%
Grade Analysis
Insoluble Matter 0.025% max. (in water)
Chemical Name or Material Acetamide
Show More Show Less
compliance-icons
Product Identifier
  • Acetamide
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.