missing translation for 'onlineSavingsMsg'
Learn More

Acetaldoxime, 99%, mixture of syn and anti

Product Code. 10162460 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10162460 100 g Glass bottle 100g
10346210 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10162460 Supplier Thermo Scientific Acros Supplier No. 102231000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 107-29-9 | C2H5NO | 59.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-29-9
Molecular Formula C2H5NO
Molecular Weight (g/mol) 59.07
MDL Number MFCD00002124 MFCD00002124
InChI Key FZENGILVLUJGJX-NSCUHMNNSA-N
Synonym acetaldoxime, z-acetaldehyde oxime, 1z-acetaldehyde oxime, acetaldehyde, oxime, 1z, 1z-ethanal oxime, nz-n-ethylidenehydroxylamine, acetaldehyde, oxime, acetaldehyde, oxime, z, z-acetaldoxime, cis-acetaldehyde oxime
PubChem CID 5324280
ChEBI CHEBI:50719
IUPAC Name (NZ)-N-ethylidenehydroxylamine
SMILES C\C=N\O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 44.0°C to 46.0°C
Color White
Density 0.9600g/mL
Boiling Point 115.0°C
Flash Point 38°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH=NOH
Refractive Index 1.4250 to 1.4270
Quantity 100 g
Beilstein 01, IV, 3121
Fieser 01,3
Merck Index 15, 43
Specific Gravity 0.96
Solubility Information Solubility in water: soluble in water. Other solubilities: decomposes by aq. hcl,soluble in alcohol and ether
Formula Weight 59.06
Percent Purity 99%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material Acetaldoxime, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetaldoxime, mixture of syn and anti
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable solid Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.