missing translation for 'onlineSavingsMsg'
Learn More

Acetaldehyde, TRC

Product Code. 30041144
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30041144 100 g 100g
18000080 1 g 1g
17919969 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30041144 Supplier Toronto Research Chemicals Supplier No. TRCA132600100G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-07-0
Molecular Formula C2 H4 O
Molecular Weight (g/mol) 44.05
Synonym Acetaldehyde (8CI, 9CI, ACI), Acetic aldehyde, Ethanal, Ethyl aldehyde, MeSH ID: D000079, NSC 7594
IUPAC Name acetaldehyde
SMILES CC=O

Specifications

Quantity 100 g
InChI Formula InChI=1S/C2H4O/c1-2-3/h2H,1H3
Formula Weight 44.0262
Chemical Name or Material Acetaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.