missing translation for 'onlineSavingsMsg'
Learn More

Acetaldehyde diethyl acetal, 99%

Product Code. 11444694
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11444694 100 g 100g
11454694 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11444694 Supplier Thermo Scientific Alfa Aesar Supplier No. A13701.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 105-57-7 | C6H14O2 | 118.18 g/mol

Acetaldehyde diethyl acetal is used as a flavoring agent to provide fruit, nut, rum, and whiskey flavors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetaldehyde diethyl acetal is used as a flavoring agent to provide fruit, nut, rum, and whiskey flavors.

Solubility
Soluble in water (46 g/L at 20°C).

Notes
Keep container tightly sealed. Store in refrigerated condition. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-57-7
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.18
MDL Number MFCD00009243
InChI Key DHKHKXVYLBGOIT-UHFFFAOYSA-N
Synonym acetal, acetaldehyde diethyl acetal, diethyl acetal, ethane, 1,1-diethoxy, diaethylacetal, acetale, ethylidene diethyl ether, acetal diethylique, diethylacetal, 1,1-dietossietano
PubChem CID 7765
IUPAC Name 1,1-diethoxyethane
SMILES CCOC(C)OCC

Specifications

Melting Point -100°C
Density 0.831
Boiling Point 101°C to 102°C
Flash Point −21°C (−6°F)
Odor Pleasant
Refractive Index 1.381
Quantity 100 g
UN Number UN1088
Beilstein 1098310
Merck Index 14,38
Solubility Information Soluble in water (46g/L at 20°C).
Formula Weight 118.18
Percent Purity 99%
Chemical Name or Material Acetaldehyde diethyl acetal
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetaldehyde diethyl acetal
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Acetal

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.